Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethyltin dichloride, 98%

Catalog No. AAA1766114
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1766114 25 g
AAA1766106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1766114 Supplier Thermo Scientific Chemicals Supplier No. A1766114
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 753-73-1 | C2H6Cl2Sn | 219.68 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753-73-1
Molecular Formula C2H6Cl2Sn
Molecular Weight (g/mol) 219.68
MDL Number MFCD00000517
InChI Key PKKGKUDPKRTKLJ-UHFFFAOYSA-L
Synonym dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicity
PubChem CID 12955
IUPAC Name dichloro(dimethyl)stannane
SMILES C[Sn](C)(Cl)Cl

Specifications

Melting Point 104°C to 108°C
Boiling Point 188°C to 190°C
Quantity 25 g
UN Number UN3146
Beilstein 3660508
Sensitivity Moisture sensitive
Formula Weight 219.69
Percent Purity 98%
Chemical Name or Material Dimethyltin dichloride
Hazard Category H301+H311-H314-H330-H360-H372
Hazard Statement GHS H Statement
H301-H311-H331-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P281-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 212-039-2
RTECSNumber WH7245000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.