Learn More
AdipoGen Dimethylvinphos
SDP
Supplier:  AdipoGen CDXD0316M050
Encompass_Preferred
Chemical. CAS 2274-67-1. Formula C10H10Cl3O4P. MW 331.5. Synthetic. Organophosphorus insecticide. Compound can be used as analytical reference material.
Catalog No. 50-256-1124