Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dimethylzinc, 1.2M solution in toluene, AcroSeal™

Catalog No. p-2799163
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC377241000 100 mL AcroSeal™ Glass bottle
AC377248000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC377241000 Supplier Thermo Scientific Chemicals Supplier No. 377241000
Only null left
Add to Cart

CAS: 544-97-8 | C2H6Zn | 95.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethylzinc
Color Clear colorless
Name Note 1.2M Solution in Toluene
CAS 544-97-8, 108-88-3
Molecular Formula C2H6Zn
Linear Formula Zn(CH3)2
MDL Number MFCD00014854
Concentration 1.2M (based on Zn) exact strength on the certificate of analysis
Density 0.9300g/mL
Merck Index 15, 3289
Synonym dimethylzinc, zinc, dimethyl, dimethyl zinc, methylzinc, unii-8h6r0n8w2f, ch3 2zn, znme2, zinc methyl, dimethylzink, hsdb 709
Solubility Information Solubility in water: vigorous reaction. Other solubilities: soluble in hydrocarbons
InChI Key JRPGMCRJPQJYPE-UHFFFAOYSA-N
SMILES [CH3-].[CH3-].[Zn+2]
IUPAC Name zinc;carbanide
Molecular Weight (g/mol) 95.45
PubChem CID 11010
Formula Weight 95.45
Packaging AcroSeal™ Glass bottle
Specific Gravity 0.93
Quantity 100 mL
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.