Learn More
Strem, An Ascensus Company CAS# 544-97-8. 50g. Dimethylzinc, min. 99% (10 wt% in hexanes) (Sure/Seal™ bottle). MFCD00014854
SDP
Catalog No. 5090111398
Encompass
CAS# 544-97-8. 50g. Dimethylzinc, min. 99% (10 wt% in hexanes) (Sure/Seal™ bottle). MFCD00014854. Molecular Weight: 95.44. Molecular Formula: Zn(CH3)2. Color/form: colorless liq. Strem# 30-5062. http://www.strem.com/catalog/v/30-5062/
Catalog No. 50-901-11398 Supplier Strem, An Ascensus Company Supplier No. 30506250G
CAS# 544-97-8. 50g. Dimethylzinc, min. 99% (10 wt% in hexanes) (Sure/Seal™ bottle). MFCD00014854. Molecular Weight: 95.44. Molecular Formula: Zn(CH3)2. Color/form: colorless liq. Strem# 30-5062. http://www.strem.com/catalog/v/30-5062/