Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethylzinc Solution

Catalog No. 41724650ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-724-650ML 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Danger
Catalog No. 41-724-650ML Supplier Sigma Aldrich Supplier No. 41724650ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethylzinc (Zn(CH3)2) is a methylating reagent used to prepare methylated organic compounds as well as organometallic compounds containing methyl groups . It is also utilized as a reagent to prepare amino alcohols, oximes, and hydrazones from arylamines, alkoxyamines, and dialkylhydrazines respectively, by radical addition reaction in the presence of air and THF.Dimethylzinc solution can be used as:A catalyst with nickel for the stereoselective C-2 alkenylation and dialkenylation of pyridine derivatives by alkynes to give monoalkenylation products.A reagent with aldehydes and 2-methoxyaniline for the synthesis of enantioselective alkyl and aralkyl secondary amines via one-pot three-component coupling reaction in the presence of zirconium tetraisopropoxide.A methylating reagent for methylation of fluoroalkylated pyruvates in the presence of copper/chiral diphosphine catalyst.

Specifications

CAS 31250-18-7
Density 0.995 g/mL at 25°C
Molecular Formula C22H36N2O6
Linear Formula C22H36N2O6
MDL Number MFCD00013324
Quantity 50 mL
Molecular Weight (g/mol) 424.53 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.