Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dimethylzinc Solution

Catalog No. 36440150ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
50 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-440-150ML 50 mL
36-440-1500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 36-440-150ML Supplier Sigma Aldrich Supplier No. 36440150ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dimethylzinc can be used as:A catalyst with nickel for the stereoselective C-2 alkenylation and dialkenylation of pyridine derivatives by alkynes to give monoalkenylation products.A reagent with aldehydes and 2-methoxyaniline for the enantioselective synthesis of alkyl and aralkyl secondary amines via one-pot three-component coupling reaction in the presence of zirconium tetraisopropoxide.A methylating reagent for the methylation of fluoroalkylated pyruvates in the presence of a copper/chiral diphosphine catalyst.An activator for radical trifluoromethylation of silyl enol ethers leading to +a-trifluoromethyl ketones.

Specifications

CAS 544-97-8
Density 0.931 g/mL at 25°C
Molecular Formula C2H6Zn
Linear Formula (CH3)2Zn
MDL Number MFCD00014854
Quantity 50 mL
Molecular Weight (g/mol) 95.46 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.