Learn More
Medchemexpress LLC Dimetridazole (1,2-dimethyl-5-nitroimidazole) | 551-92-8 | 99.8% | 500 MG
SDP
Catalog No. 5000411592
Encompass_Preferred
Dimetridazole is a nitroimidazole antibiotic that inhibits protein synthesis in cultures of *Campylobacter jejuni*. It is genotoxic and can be utilized in the research of protozoal and bacterial infections.
  • Inhibits protein synthesis in *Campylobacter jejuni* cultures.
  • Shows activity against *Bacteroides fragilis* isolates.
  • Induces dose-dependent DNA damage in human lymphocytes.
  • Useful in research of protozoal infections.
  • Useful in research of bacterial infections.
Catalog No. 50-004-11592 Supplier Medchemexpress LLC Supplier No. HYB1244500MG
Dimetridazole is a nitroimidazole antibiotic that inhibits protein synthesis in cultures of *Campylobacter jejuni*. It is genotoxic and can be utilized in the research of protozoal and bacterial infections.
  • Inhibits protein synthesis in *Campylobacter jejuni* cultures.
  • Shows activity against *Bacteroides fragilis* isolates.
  • Induces dose-dependent DNA damage in human lymphocytes.
  • Useful in research of protozoal infections.
  • Useful in research of bacterial infections.