Learn More
Medchemexpress LLC Dimetridazole-d3 (1,2-Dimethyl-5-nitroimidazole-d3) | 64678-69-9 | 99.8% | 1 MG
SDP
Catalog No. 5000413113
Encompass_Preferred
Dimetridazole-d3 (1,2-Dimethyl-5-nitroimidazole-d3) is a deuterium-labeled version of Dimetridazole, a nitroimidazole antibiotic that inhibits protein synthesis in Campylobacter jejuni cultures and has genotoxic properties. This compound is suitable for research on protozoal and bacterial infections.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-13113 Supplier Medchemexpress LLC Supplier No. HYB1244S1MG
Dimetridazole-d3 (1,2-Dimethyl-5-nitroimidazole-d3) is a deuterium-labeled version of Dimetridazole, a nitroimidazole antibiotic that inhibits protein synthesis in Campylobacter jejuni cultures and has genotoxic properties. This compound is suitable for research on protozoal and bacterial infections.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can influence the pharmacokinetic and metabolic profiles of drugs.