Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimetridazole-d3 (1,2-dimethyl-5-nitroimidazole-d3) | 64678-69-9 | 99.8% | 5 MG
SDP

Catalog No. 5000412920
Encompass_Preferred

Dimetridazole-d3 is a deuterium labeled nitroimidazole antibiotic that inhibits protein synthesis in cultures of Campylobacter jejuni and is genotoxic. It is used in the research of protozoal and bacterial infections, and can serve as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.

  • Deuterium labeled
  • Nitroimidazole antibiotic
  • Inhibits protein synthesis in cultures of Campylobacter jejuni
  • Genotoxic
  • Suitable for research of protozoal and bacterial infections
  • Functions as a tracer
  • Functions as an internal standard for quantitative analysis
  • Molecular weight: 144.15
  • Formula: C5H4D3N3O2
  • Appearance: solid
  • Color: white to off-white
  • Solubility in DMSO: 50 mg/mL

Catalog No. 50-004-12920 Supplier Medchemexpress LLC Supplier No. HYB1244S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimetridazole-d3 is a deuterium labeled nitroimidazole antibiotic that inhibits protein synthesis in cultures of Campylobacter jejuni and is genotoxic. It is used in the research of protozoal and bacterial infections, and can serve as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.

  • Deuterium labeled
  • Nitroimidazole antibiotic
  • Inhibits protein synthesis in cultures of Campylobacter jejuni
  • Genotoxic
  • Suitable for research of protozoal and bacterial infections
  • Functions as a tracer
  • Functions as an internal standard for quantitative analysis
  • Molecular weight: 144.15
  • Formula: C5H4D3N3O2
  • Appearance: solid
  • Color: white to off-white
  • Solubility in DMSO: 50 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.