Learn More
Cayman Chemical DImetrIdazole-d3 1mg
SDP
Supplier:  Cayman Chemical 278411 MG
Encompass_Preferred
An internal standard for the quantification of dimetridazole by GC- or LC-MS
Catalog No. 50-294-0414