Learn More
Cayman Chemical DImetrIdazole-d3 500ug
SDP
Catalog No. 502938588
Encompass_Preferred
An internal standard for the quantification of dimetridazole by GC- or LC-MS
Catalog No. 50-293-8588 Supplier Cayman Chemical Supplier No. 27841500 UG
An internal standard for the quantification of dimetridazole by GC- or LC-MS