Learn More
AdipoGen Dimidium bromide
SDP
Catalog No. 502561267
Encompass_Preferred
Chemical. CAS 518-67-2. Formula C20H18BrN3. MW 380.28. Synthetic. Intercalating probe for nucleic acids. Other applications include the determination of cationic surfactants.
Catalog No. 50-256-1267 Supplier AdipoGen Supplier No. CDXD0065M250
Chemical. CAS 518-67-2. Formula C20H18BrN3. MW 380.28. Synthetic. Intercalating probe for nucleic acids. Other applications include the determination of cationic surfactants.