Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimidium bromide | 518-67-2 | 98.6% | 1 G
SDP

Catalog No. 5000410932
Encompass_Preferred

Dimidium bromide is a biochemical reagent used in life science research as a biological material or organic compound.

  • Solid appearance
  • Pale purple to purple color
  • Store at room temperature, sealed, away from moisture and light
  • In solvent, store at -80°C for 1 year or -20°C for 6 months
  • Soluble in DMSO (125 mg/mL)

Catalog No. 50-004-10932 Supplier Medchemexpress LLC Supplier No. HYD07881G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimidium bromide is a biochemical reagent used in life science research as a biological material or organic compound.

  • Solid appearance
  • Pale purple to purple color
  • Store at room temperature, sealed, away from moisture and light
  • In solvent, store at -80°C for 1 year or -20°C for 6 months
  • Soluble in DMSO (125 mg/mL)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.