Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dimidium bromide | 518-67-2 | 98.6% | 5 G
SDP

Catalog No. 5000410933
Encompass_Preferred

Dimidium bromide is a biochemical reagent suitable for use as a biological material or organic compound in life science-related research. It is intended for research use only and is not sold to patients.

  • Molecular weight: 380.28
  • Formula: C20H18BrN3
  • Appearance: Solid
  • Color: Pale purple to purple
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: RT, sealed storage, away from moisture and light. In solvent: -80°C for 1 year, -20°C for 6 months
  • Solubility (in vitro): DMSO : 125 mg/mL (328.71 mM; requires ultrasonic; hygroscopic DMSO significantly impacts solubility, use newly opened DMSO)

Catalog No. 50-004-10933 Supplier Medchemexpress LLC Supplier No. HYD07885G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Dimidium bromide is a biochemical reagent suitable for use as a biological material or organic compound in life science-related research. It is intended for research use only and is not sold to patients.

  • Molecular weight: 380.28
  • Formula: C20H18BrN3
  • Appearance: Solid
  • Color: Pale purple to purple
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: RT, sealed storage, away from moisture and light. In solvent: -80°C for 1 year, -20°C for 6 months
  • Solubility (in vitro): DMSO : 125 mg/mL (328.71 mM; requires ultrasonic; hygroscopic DMSO significantly impacts solubility, use newly opened DMSO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.