Learn More
Selleck Chemical LLC Diminazene Aceturate-50mg
SDP
Catalog No. 502389987
Encompass_Preferred
Diminazene(Diminazene diaceturate) is a di-amidine also known as 4,4-(1-Triazene–1,3–diyl)bis(benzenecarboximidamide), used as an effective trypanocidal agent. The main biochemical mechanism of the trypanocidal actions of Diminazene aceturate is by binding to trypanosomal kinetoplast DNA (kDNA) in a non-intercalative manner through specific interaction with sites rich in adenine-thymine base pairs. Diminazene aceturate is also an angiotensin-converting enzyme 2 (ACE2) activator and has strong and potent anti-inflammatory properties.
Catalog No. 50-238-9987 Supplier Selleck Chemical LLC Supplier No. S410450MG
Diminazene(Diminazene diaceturate) is a di-amidine also known as 4,4-(1-Triazene–1,3–diyl)bis(benzenecarboximidamide), used as an effective trypanocidal agent. The main biochemical mechanism of the trypanocidal actions of Diminazene aceturate is by binding to trypanosomal kinetoplast DNA (kDNA) in a non-intercalative manner through specific interaction with sites rich in adenine-thymine base pairs. Diminazene aceturate is also an angiotensin-converting enzyme 2 (ACE2) activator and has strong and potent anti-inflammatory properties.