Learn More
Lkt Laboratories Diminazene Aceturate, ≥98%, 1GR
SDP
Supplier:  Lkt Laboratories D33531G
Encompass_Preferred
Diamidine; ACE2 activator, DNA polymerase inhibitor. CAS: 908-54-3. Additional sizes available upon request. Ships from Saint Paul, MN.
Catalog No. 50-153-4903