Learn More
Alkali Scientific Dimpled Mat, Large 12" X 12", for BenchRocker™ 3D Nutating Shaker , 1/pk
SDP
Catalog No. 502388658
Encompass_Preferred
Dimpled Mat, large 12" x 12". Compatible with BenchBlotter™ Platform Rocker, BenchBlotter™ 2D Rocker, Everlast™ Rocker, MiniMixer™ 3D Nutating Shaker, BioMixer™ 3D Nutating Shaker, Mini BlotBoy™ 3D Rocker, BlotBoy™ 3D Rocker, and BenchRocker™ 3D Nutating Shaker.
Catalog No. 50-238-8658 Supplier Alkali Scientific Supplier No. RS70043
Dimpled Mat, large 12" x 12". Compatible with BenchBlotter™ Platform Rocker, BenchBlotter™ 2D Rocker, Everlast™ Rocker, MiniMixer™ 3D Nutating Shaker, BioMixer™ 3D Nutating Shaker, Mini BlotBoy™ 3D Rocker, BlotBoy™ 3D Rocker, and BenchRocker™ 3D Nutating Shaker.