Learn More
Medchemexpress LLC HY-10492 10mg Medchemexpress, Dinaciclib CAS:779353-01-4 Purity:>98%
SDP
Catalog No. 502028761
Encompass_Preferred
Medchemexpress, HY-10492 10mg Dinaciclib CAS:779353-01-4 Dinaciclib (SCH 727965) is a potent inhibitor of CDK, with IC50s of 1 nM, 1 nM, 3 nM, and 4 nM for CDK2, CDK5, CDK1, and CDK9, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8761 Supplier Medchemexpress LLC Supplier No. HY1049210MG
Medchemexpress, HY-10492 10mg Dinaciclib CAS:779353-01-4 Dinaciclib (SCH 727965) is a potent inhibitor of CDK, with IC50s of 1 nM, 1 nM, 3 nM, and 4 nM for CDK2, CDK5, CDK1, and CDK9, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.