Learn More
TARGETMOL CHEMICALS INC DINACICLIB 10MG
SDP
Catalog No. 502577277
Encompass_Preferred
Also available in 2 mg 5 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dinaciclib (SCH 727965) is a new-type and effective CDK inhibitor for CDK2/5/1/9 (IC50 1 nM/1 nM/3 nM/4 nM) with potential antineoplastic activity. purity: 99%
Catalog No. 50-257-7277 Supplier TARGETMOL CHEMICALS INC Supplier No. T191210MG
Add to Cart
Add to Cart
Also available in 2 mg 5 mg 50 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dinaciclib (SCH 727965) is a new-type and effective CDK inhibitor for CDK2/5/1/9 (IC50 1 nM/1 nM/3 nM/4 nM) with potential antineoplastic activity. purity: 99%