Learn More
Selleck Chemical LLC Dinaciclib S2768-200mg
SDP
Catalog No. 503112030
Encompass_Preferred
Dinaciclib is a novel and potent CDK inhibitor for CDK2 CDK5 CDK1 and CDK9 with IC50 of 1 nM 1 nM 3 nM and 4 nM in cell-free assays respectively It also blocks thymidine (dThd) DNA incorporation Dinaciclib induces apoptosis through the activation of caspases 8 and 9 Phase 3
Catalog No. 50-311-2030 Supplier Selleck Chemical LLC Supplier No. S2768200MG
Dinaciclib is a novel and potent CDK inhibitor for CDK2 CDK5 CDK1 and CDK9 with IC50 of 1 nM 1 nM 3 nM and 4 nM in cell-free assays respectively It also blocks thymidine (dThd) DNA incorporation Dinaciclib induces apoptosis through the activation of caspases 8 and 9 Phase 3