Learn More
Medchemexpress LLC Dinaciclib 200mg | 779353-01-4 | 200MG
SDP
Catalog No. 5000243306
Encompass_Preferred
Dinaciclib (SCH 727965) is a potent inhibitor of CDK with IC50s of 1 nM 1 nM 3 nM and 4 nM for CDK2 CDK5 CDK1 and CDK9 respectively[1
Catalog No. 50-002-43306 Supplier Medchemexpress LLC Supplier No. HY10492200MG
Dinaciclib (SCH 727965) is a potent inhibitor of CDK with IC50s of 1 nM 1 nM 3 nM and 4 nM for CDK2 CDK5 CDK1 and CDK9 respectively[1