Learn More
Abcam Dinaciclib, cyclin-dependent kinase (CDK) inhibitor, 5MG
Catalog No. 502672803
Encompass_Preferred
MW 396.5 Da, Purity >98%. Novel, potent, selective cyclin-dependent kinase (CDK) inhibitor displaying selectivity for CDK1, CDK2, CDK5 and CDK9 (IC₅₀ values are 3, 1, 1 and 4 nM respectively). Potent DNA replication inhibitor that blocks thymidine incorporation in A2780 ovarian cancer cells (IC₅₀ = 4 nM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-2803 Supplier Abcam Supplier No. AB2194695MG
MW 396.5 Da, Purity >98%. Novel, potent, selective cyclin-dependent kinase (CDK) inhibitor displaying selectivity for CDK1, CDK2, CDK5 and CDK9 (IC₅₀ values are 3, 1, 1 and 4 nM respectively). Potent DNA replication inhibitor that blocks thymidine incorporation in A2780 ovarian cancer cells (IC₅₀ = 4 nM).
The product is subject to the following: Abcam Restricted Use Statement