Learn More
Selleck Chemical LLC Dinaciclib (SCH727965) 5mg 779353-01-4 PS-095760
SDP
Supplier:  Selleck Chemical LLC S27685MG
Encompass_Preferred
Dinaciclib (SCH727965, PS-095760) is a novel and potent CDK inhibitor for CDK2, CDK5, CDK1 and CDK9 with IC50 of 1 nM, 1 nM, 3 nM and 4 nM in cell-free assays, respectively. It also blocks thymidine (dThd) DNA incorporation. Dinaciclib induces apoptosis through the activation of caspases 8 and 9. Phase 3. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0666375