Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Dinaphtho[2,3-b:2',3'-f]thieno[3,2-b]thiophene

Catalog No. 767638100MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-763-8100MG 100 mg
76-763-8500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 76-763-8100MG Supplier Sigma Aldrich Supplier No. 767638100MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Dinaphtho[2,3-b:2',3'-f]thieno[3,2-b]thiophene

Specifications

CAS 935280-42-5
Melting Point 425°C to 430°C
Molecular Formula C22H12S2
Linear Formula C22H12S2
Quantity 100 mg
MDL Number MFCD22666453
Synonym DNTT; Naphtho[2,3-b]naphtho[2 ,3 :4,5 ]thieno[2,3-d]thiophene
Molecular Weight (g/mol) 340.46
Percent Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.