Learn More
Cayman Chemical Dianhydrogalactitol, Purity: ≥95%, CAS: 23261-20-3, 10 mg
SDP
Catalog No. 502917796
Encompass_Preferred
A DNA alkylating agent; induces N7-alkylation of guanine and interstrand DNA cross-links in vitro; halts the cell cycle at the G2/M phase in U-251MG and T98G cells at 0.5-8 µM; decreases the migration and invasion of U251 cells and tube formation of HUVECs at 12.5 µg/ml; increases survival in a U-251MG mouse xenograft model when administered in combination with temozolomide
Catalog No. 50-291-7796 Supplier Cayman Chemical Supplier No. 3770510 MG
A DNA alkylating agent; induces N7-alkylation of guanine and interstrand DNA cross-links in vitro; halts the cell cycle at the G2/M phase in U-251MG and T98G cells at 0.5-8 µM; decreases the migration and invasion of U251 cells and tube formation of HUVECs at 12.5 µg/ml; increases survival in a U-251MG mouse xenograft model when administered in combination with temozolomide