Learn More
Cayman Chemical Dinitolmide, Purity: ≥98%, CAS: 148-01-6, 100 mg
SDP
Catalog No. 502989274
Encompass_Preferred
An anticoccidial agent; dietary administration reduces mortality and decreases the number of oocysts and oocyst sporulation in chick models of E. maxima, E. brunetti, or E. acervulina infection from 125-500 ppm
Catalog No. 50-298-9274 Supplier Cayman Chemical Supplier No. 31138100 MG
An anticoccidial agent; dietary administration reduces mortality and decreases the number of oocysts and oocyst sporulation in chick models of E. maxima, E. brunetti, or E. acervulina infection from 125-500 ppm