Learn More
Medchemexpress LLC Dinitolmide | 148-01-6 | 99.7% | 250 MG
SDP
Catalog No. 5000406572
Encompass_Preferred
Dinitolmide (Zoalene) is an orally active anticoccidial agent used as a fodder additive. It has a wide anticoccidial spectrum and does not affect host immunity. It primarily acts on the first-generation merozoite stage of coccidia and also inhibits the sporulation process of oocysts. This compound exhibits anti-Toxoplasma gondii effects in vitro and can inhibit infections caused by *Eimeria tenella*.
  • Orally active anticoccidial agent
  • Used as a fodder additive
  • Possesses a wide anticoccidial spectrum
  • Does not affect host immunity
  • Acts on the first-generation merozoite stage of coccidia
  • Inhibits oocyst sporulation
  • Exhibits anti-Toxoplasma gondii effects
  • Inhibits *Eimeria tenella* infections
Catalog No. 50-004-06572 Supplier Medchemexpress LLC Supplier No. 250MGHYB1004
Add to Cart
Add to Cart
Dinitolmide (Zoalene) is an orally active anticoccidial agent used as a fodder additive. It has a wide anticoccidial spectrum and does not affect host immunity. It primarily acts on the first-generation merozoite stage of coccidia and also inhibits the sporulation process of oocysts. This compound exhibits anti-Toxoplasma gondii effects in vitro and can inhibit infections caused by *Eimeria tenella*.
  • Orally active anticoccidial agent
  • Used as a fodder additive
  • Possesses a wide anticoccidial spectrum
  • Does not affect host immunity
  • Acts on the first-generation merozoite stage of coccidia
  • Inhibits oocyst sporulation
  • Exhibits anti-Toxoplasma gondii effects
  • Inhibits *Eimeria tenella* infections