Learn More
Cayman Chemical DInItolmIde 50mg
SDP
Catalog No. 502989296
Encompass_Preferred
An anticoccidial agent; dietary administration reduces mortality and decreases the number of oocysts and oocyst sporulation in chick models of E. maxima, E. brunetti, or E. acervulina infection from 125-500 ppm
Catalog No. 50-298-9296 Supplier Cayman Chemical Supplier No. 3113850 MG
An anticoccidial agent; dietary administration reduces mortality and decreases the number of oocysts and oocyst sporulation in chick models of E. maxima, E. brunetti, or E. acervulina infection from 125-500 ppm