Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dinitronaphthalene (mixture) 85.0+%, TCI America™
SDP

Catalog No. D083825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D083825G 25 g
D0838500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D083825G Supplier TCI America Supplier No. D083825G

Chemical Identifiers

CAS 27478-34-8
Molecular Formula C10H6N2O4
Molecular Weight (g/mol) 218.168
MDL Number MFCD01778365
InChI Key XNKFCDGEFCOQOM-UHFFFAOYSA-N
PubChem CID 612684
IUPAC Name 1,2-dinitronaphthalene
SMILES C1=CC=C2C(=C1)C=CC(=C2[N+](=O)[O-])[N+](=O)[O-]

Specifications

Quantity 25 g
Formula Weight 218.17
Percent Purity ≥85.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Dinitronaphthalene (mixture)
EINECSNumber (4)-0325
RTECSNumber QJ4550000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.