Learn More
Dino Lite USB Eyepiece Camera AM7025X – 5MP, Use on Traditional Microscope
Catalog No. NC1486986
Encompass_Preferred
The 5MP Dino-Eye Edge Series AM7025X offers remarkable image performance while providing a relatively larger field of view than its predecessors. AM7025X design and included adapters allows for fitment within 23mm, 30mm, 30.5mm or c-mount eyepiece slots which can be fixed into ideal focal positions.
Catalog No. NC1486986 Supplier Dino Lite Supplier No. AM7025X
The 5MP Dino-Eye Edge Series AM7025X offers remarkable image performance while providing a relatively larger field of view than its predecessors. AM7025X design and included adapters allows for fitment within 23mm, 30mm, 30.5mm or c-mount eyepiece slots which can be fixed into ideal focal positions.