Learn More
Dino Lite MS33A Adjustable Articulating Arm - Designed for Dino-Lite
Catalog No. NC1678714
Encompass_Preferred
MS33A is a fully adjustable articulating arm designed for Dino-Lite microscope cameras. The articulating arm can be handled and adjusted simply with one hand. Tighten the single knob locking mechanism to hold the Dino-Lite in the desired position. When extended flat, the arm has a 9" reach.
Catalog No. NC1678714 Supplier Dino Lite Supplier No. MS33A
MS33A is a fully adjustable articulating arm designed for Dino-Lite microscope cameras. The articulating arm can be handled and adjusted simply with one hand. Tighten the single knob locking mechanism to hold the Dino-Lite in the desired position. When extended flat, the arm has a 9" reach.