Learn More
Dino Lite USB 3.0 Digital Microscope AM73115MTF - 5MP, 10x - 70x Optical Magnification, Measurement, Far Working Distance
Catalog No. NC1678713
Encompass_Preferred
Dino-Lite Edge AM73115MTF is a 'far' working distance digital microscope that utilizes USB 3.0. It provides uncompressed viewing to deliver exceptional sharpness, improved color accuracy and image quality. AM73115MTF is designed to provide the farthest available working distance with a magnification range of 10x - 70x. By using USB 3.0, the AM73115MTF is capable of providing 45FPS at 1280 x 960 resolution. The far working distance impressively offers minimum 115mm viewing distance with maximum 70x magnification.Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. The software also allows users to disable the MicroTouch, or set to single or double touch to trigger.
Catalog No. NC1678713 Supplier Dino Lite Supplier No. AM73115MTF
Dino-Lite Edge AM73115MTF is a 'far' working distance digital microscope that utilizes USB 3.0. It provides uncompressed viewing to deliver exceptional sharpness, improved color accuracy and image quality. AM73115MTF is designed to provide the farthest available working distance with a magnification range of 10x - 70x. By using USB 3.0, the AM73115MTF is capable of providing 45FPS at 1280 x 960 resolution. The far working distance impressively offers minimum 115mm viewing distance with maximum 70x magnification.Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. The software also allows users to disable the MicroTouch, or set to single or double touch to trigger.