Learn More
Dino Lite USB 3.0 Digital Microscope AM73915MZT - 5MP, 10x - 220x Optical Magnification, Measurement, Polarized Light, AMR, EDOF
Catalog No. NC1768534
Encompass_Preferred
Dino-Lite Edge AM73915MZT provides uncompressed viewing to deliver exceptional sharpness, improved color accuracy and image quality. It offers a a magnification range of 10x - 220x. By using USB 3.0, the AM73915MZT is capable of providing 45FPS at 1280 x 960 resolution, allowing this microscope to be used in a wider range of more demanding applications. Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. Polarization can be switched on/off or can be adjusted to offer full or half polarization for controlling glare. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. Flexible LED Control (FLC) enables partial illumination control options to enhance edge contrast and provides intensity adjustment for better adaptability, combined with the built-on polarizer, interchangeable front caps, and rugged metal housing.
Catalog No. NC1768534 Supplier Dino Lite Supplier No. AM73915MZT
Dino-Lite Edge AM73915MZT provides uncompressed viewing to deliver exceptional sharpness, improved color accuracy and image quality. It offers a a magnification range of 10x - 220x. By using USB 3.0, the AM73915MZT is capable of providing 45FPS at 1280 x 960 resolution, allowing this microscope to be used in a wider range of more demanding applications. Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. Polarization can be switched on/off or can be adjusted to offer full or half polarization for controlling glare. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. Flexible LED Control (FLC) enables partial illumination control options to enhance edge contrast and provides intensity adjustment for better adaptability, combined with the built-on polarizer, interchangeable front caps, and rugged metal housing.