Learn More
Dino Lite RK-06-PL Extra Extension Pole for RK-06A
Catalog No. NC1636214
Encompass_Preferred
RK-06-PL is an optional accessory extension pole to double the height of the RK-06A stand.
Catalog No. NC1636214 Supplier Dino Lite Supplier No. RK06PL
RK-06-PL is an optional accessory extension pole to double the height of the RK-06A stand.