Learn More
Dino Lite RCA Digital Microscope AM4112NT – 640 x 480 Resolution, 10x - 50x, 220x Optical Magnification
Catalog No. NC1835638
Encompass_Preferred
The AM4112NT Dino-Lite handheld microscope is a composite model that connects directly to a television. It has AV/Video RCA output, which connects to the TV through the RCA/Component cables. This model will excel with real-time imaging and viewing objects in motion. With no software needed, this makes quick inspection more efficient than before. This model uses a magnification dial for smooth and easy adjustment and focus. This model also has a scroll lock feature to keep the desired set magnification/focus. Capture in full detail with resolution up to 640 x 480 pixels.
Catalog No. NC1835638 Supplier Dino Lite Supplier No. AM4112NT
The AM4112NT Dino-Lite handheld microscope is a composite model that connects directly to a television. It has AV/Video RCA output, which connects to the TV through the RCA/Component cables. This model will excel with real-time imaging and viewing objects in motion. With no software needed, this makes quick inspection more efficient than before. This model uses a magnification dial for smooth and easy adjustment and focus. This model also has a scroll lock feature to keep the desired set magnification/focus. Capture in full detail with resolution up to 640 x 480 pixels.