Learn More
Dino Lite MS35B Rigid Tabletop Stand Designed for Dino-Lite
Catalog No. NC1272981
Encompass_Preferred
The MS35B is a versatile, lightweight and economical vertical stand with fine adjustment designed for use with the Dino-Lite handheld digital microscope cameras. It has a range of up to 10 inches of vertical working distance from the object. The main body assembly with slides up or down into position on the vertical post and can be rotated 360 degrees, then secured into position with a rear set screw. The Dino-Lite handheld microscope (not included) slides directly into a holster which is mounted onto a peg located on the main body.
Catalog No. NC1272981 Supplier Dino Lite Supplier No. MS35B
Add to Cart
Add to Cart
The MS35B is a versatile, lightweight and economical vertical stand with fine adjustment designed for use with the Dino-Lite handheld digital microscope cameras. It has a range of up to 10 inches of vertical working distance from the object. The main body assembly with slides up or down into position on the vertical post and can be rotated 360 degrees, then secured into position with a rear set screw. The Dino-Lite handheld microscope (not included) slides directly into a holster which is mounted onto a peg located on the main body.