Learn More
Dino Lite BL-ZW1 Polarized Back-light Stage
Catalog No. NC1965402
Encompass_Preferred
BL-ZW1 is a backlight stage polarizer designed for use with Dino-Lite models with built on polarizers. (Examples: AM4113ZT, AF4115ZT, AF4915ZT, AM7115ZT, etc). BL-ZW1 is a sturdy and attractive backlight stage that is powered through a USB cord or AC Adapter. Adjustment of the polarizer is simply performed by sliding the wheel adjustment located on the front of the stage. Diameter of illumination: 4.5cm.
Catalog No. NC1965402 Supplier Dino Lite Supplier No. BLZW1
BL-ZW1 is a backlight stage polarizer designed for use with Dino-Lite models with built on polarizers. (Examples: AM4113ZT, AF4115ZT, AF4915ZT, AM7115ZT, etc). BL-ZW1 is a sturdy and attractive backlight stage that is powered through a USB cord or AC Adapter. Adjustment of the polarizer is simply performed by sliding the wheel adjustment located on the front of the stage. Diameter of illumination: 4.5cm.