Learn More
Dino Lite USB Digital Microscope AF3113T - 0.3MP, 20x - 220x Optical Magnification, Measurement, WF-20 Compatible
Catalog No. NC1864214
Encompass_Preferred
AF3113T is designed to meet most applications within 20x- 220x magnification with 0.3MP. The new modular wireless-ready design allows for USB connection or a quick swap to wireless with a WF-20 adapter (sold separately) making it a completely cordless and portable microscope. AF3113T has measurement features within the included software. Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. The software also allows users to disable the MicroTouch, or set to single or double touch to trigger. This model is wireless-ready, but requires the WF-20 adapter for wireless functionality.
Catalog No. NC1864214 Supplier Dino Lite Supplier No. AF3113T GRAY
AF3113T is designed to meet most applications within 20x- 220x magnification with 0.3MP. The new modular wireless-ready design allows for USB connection or a quick swap to wireless with a WF-20 adapter (sold separately) making it a completely cordless and portable microscope. AF3113T has measurement features within the included software. Measurements can be performed in different units (inch/mil/mm/um) and types including straight lines, circles, angles, arches grids and more. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. The software also allows users to disable the MicroTouch, or set to single or double touch to trigger. This model is wireless-ready, but requires the WF-20 adapter for wireless functionality.