Learn More
Microscope Store DINO-LITE TABLE STAND
SDP
Catalog No. NC0638773
Encompass_Preferred
The most popular industrial inspection stand with 8 in. max WD, 360 deg. positioning, +/- 40 deg. viewing angles. Vernier control for precise focusing. Stainless steeel/aluminum construction. Can be paired with MS15X rotating stage.
Catalog No. NC0638773 Supplier Microscope Store Supplier No. MS35B
The most popular industrial inspection stand with 8 in. max WD, 360 deg. positioning, +/- 40 deg. viewing angles. Vernier control for precise focusing. Stainless steeel/aluminum construction. Can be paired with MS15X rotating stage.