Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dino Math Tracks™ Place Value Game
SCI_ED

Catalog No. s79004
Change view
Click to view available options
:
Dino Math Tracks
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S79004 Dino Math Tracks
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S79004 Supplier Learning Resources, Inc Supplier No. LER 0712
Only null left
Add to Cart
Add to Cart

Includes

game board, 16 dinosaurs, cards, number dice, and instructions for various levels of play.

The way to make math fun

Roll the dice to move dinosaurs around the board and to reinforce counting, addition, subtraction, and place value skills. For 2 to 4 players.

Specifications

For Use With (Application) Reinforce counting, addition, subtraction, and place value skills
Includes Game board, 16 dinosaurs, cards, number dice, and instructions for various levels of play
Class Size 2 to 4 Students
Description Dino Math Tracks™ Place Value Game
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.