Learn More
Dino Lite Dino-Lite 90 Degree Mirror View Adapter
Catalog No. NC3373884
Encompass_Preferred
Attachable 90 degree view adapter that is compatible with Dino-Lite “AD” series extended working distance, “L” models Compatible with Dino-Lite "AD" series long working distance, "L" models: AD4113TL, AD4112NTL
Catalog No. NC3373884 Supplier Dino Lite Supplier No. FCMA1
Attachable 90 degree view adapter that is compatible with Dino-Lite “AD” series extended working distance, “L” models Compatible with Dino-Lite "AD" series long working distance, "L" models: AD4113TL, AD4112NTL