Learn More
Dino Lite USB Digital Microscope AM4111T - 1.3MP, 10x - 50x, 220x Optical Magnification, MicroTouch
Catalog No. NC1420073
Encompass_Preferred
The AM4111T handheld digital microscope comes with a high quality 1.3 megapixel image sensor that views and takes highly detailed images but do not require measurement. This model is featured with the unique MicroTouch snapshot feature. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. The software also allows users to disable the MicroTouch, or set to single or double touch to trigger. Capture in full detail with resolution up to 1280 x 1024 pixels. Use simple tools to draw lines, shapes, and text annotations on the image.
Catalog No. NC1420073 Supplier Dino Lite Supplier No. AM4111T
The AM4111T handheld digital microscope comes with a high quality 1.3 megapixel image sensor that views and takes highly detailed images but do not require measurement. This model is featured with the unique MicroTouch snapshot feature. The MicroTouch feature is a touch sensitive area which can be used to capture images instead of using a keyboard and mouse. The software also allows users to disable the MicroTouch, or set to single or double touch to trigger. Capture in full detail with resolution up to 1280 x 1024 pixels. Use simple tools to draw lines, shapes, and text annotations on the image.