Learn More
AdipoGen Dinotefuran
SDP
Supplier:  AdipoGen CDXD0309G001
Encompass_Preferred
Chemical. CAS 165252-70-0. Formula C7H14N4O3. MW 202.2. Synthetic. A neonicotinoid insecticide. Compound can be used as analytical reference material.
Catalog No. 50-256-1132