Learn More
Medchemexpress LLC Dinotefuran-d3 | 2934813-27-9 | 98.71% | 1 MG
SDP
Catalog No. 5000408822
Encompass_Preferred
Dinotefuran-d3 is a deuterium-labeled variant of Dinotefuran, an insecticide belonging to the neonicotinoid class. Its primary mode of action is to disrupt the insect's nervous system by inhibiting nicotinic acetylcholine receptors. This product is exclusively for research purposes and is not for sale to patients.
  • Serves as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution may influence the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-08822 Supplier Medchemexpress LLC Supplier No. HYB0827S1MG
Dinotefuran-d3 is a deuterium-labeled variant of Dinotefuran, an insecticide belonging to the neonicotinoid class. Its primary mode of action is to disrupt the insect's nervous system by inhibiting nicotinic acetylcholine receptors. This product is exclusively for research purposes and is not for sale to patients.
  • Serves as a tracer for quantitation during drug development.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium substitution may influence the pharmacokinetic and metabolic profiles of drugs.