Learn More
Biotium DIOC14(3)
Catalog No. 501964717
Encompass_Preferred
DiOC14(3) is a DiO derivative, but is more soluble in aqueous media than the latter because it has shorter hydrocarbon chains. Also offered as ready-to-use CellBrite™ Cell Membrane Labeling Solutions. Properties: Ex/ Em(MeOH) = 484/501nm; e (MeOH) = 150,000; Orange solid soluble in methanol, ethanol or DMSO; Store desiccated at 4°C; C47H74N2O6S; MW: 795
Catalog No. 50-196-4717 Supplier Biotium Supplier No. 60012
DiOC14(3) is a DiO derivative, but is more soluble in aqueous media than the latter because it has shorter hydrocarbon chains. Also offered as ready-to-use CellBrite™ Cell Membrane Labeling Solutions. Properties: Ex/ Em(MeOH) = 484/501nm; e (MeOH) = 150,000; Orange solid soluble in methanol, ethanol or DMSO; Store desiccated at 4°C; C47H74N2O6S; MW: 795