Learn More
Biotium DiOC5(3)
Catalog No. 501964743
Encompass_Preferred
DiOC5(3) and DiOC6(3) are two of the most widely used carbocyanine dyes used for membrane potential measurements.Properties: Ex/Em(MeOH) = 482/497 nm; Orange solid soluble in DMSO or DMF; Store at 4°C and protect from light; C27H33IN2O2; MW: 544.47; [53213-81-3 ];
Catalog No. 50-196-4743 Supplier Biotium Supplier No. 70007
DiOC5(3) and DiOC6(3) are two of the most widely used carbocyanine dyes used for membrane potential measurements.Properties: Ex/Em(MeOH) = 482/497 nm; Orange solid soluble in DMSO or DMF; Store at 4°C and protect from light; C27H33IN2O2; MW: 544.47; [53213-81-3 ];