Learn More
STEMCELL Technologies DiOC63 Iodide
SDP
Catalog No. 502740896
Encompass
Green fluorescent lipophilic dye for staining cell membranes and other hydrophobic structures in live and fixed cells - 25 Mg
Catalog No. 50-274-0896 Supplier STEMCELL Technologies Supplier No. 1000815
Green fluorescent lipophilic dye for staining cell membranes and other hydrophobic structures in live and fixed cells - 25 Mg