Learn More
Abcam DiD, 10MG
Supplier:  Abcam AB27531910MG
Encompass_Preferred
MW 959.9 Da. Red fluorescent membrane probe for long term staining of live cells, including neuronal tracing -due to the long chain (C-18) groups that embed into the plasma membrane of live cells. Also used in fiber optic sensors for detecting K+ using fluorescence energy transfer.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-996-238