Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dioctadecyl 3,3'-Thiodipropionate 90.0+%, TCI America™
SDP

Catalog No. T020625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T020625G 25 g
T0206500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T020625G Supplier TCI America Supplier No. T020625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 693-36-7
Molecular Formula C42H82O4S
Molecular Weight (g/mol) 683.174
MDL Number MFCD00026684
InChI Key PWWSSIYVTQUJQQ-UHFFFAOYSA-N
PubChem CID 12738
IUPAC Name octadecyl 3-(3-octadecoxy-3-oxopropyl)sulfanylpropanoate
SMILES CCCCCCCCCCCCCCCCCCOC(=O)CCSCCC(=O)OCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 65°C
Color White
Quantity 25 g
Synonym Distearyl 3,3′C-Thiodipropionate, 3,3′C-Thiodipropionic Acid Dioctadecyl Ester
Formula Weight 683.17
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dioctadecyl 3,3′-Thiodipropionate
EINECSNumber (2)-0919&(2)-1399
RTECSNumber UF8010000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.