Learn More
Cayman Chemical Dioctanoyl Glycol, Purity: ≥98%, CAS: 627-86-1, 50 mg
SDP
Catalog No. 502990675
Encompass_Preferred
A DGK inhibitor (Ki = 58 μM); reduces basal and thrombin-stimulated phosphatidic acid levels in washed isolated human platelets at 30, 100, and 500 μM
Catalog No. 50-299-0675 Supplier Cayman Chemical Supplier No. 3151550 MG
A DGK inhibitor (Ki = 58 μM); reduces basal and thrombin-stimulated phosphatidic acid levels in washed isolated human platelets at 30, 100, and 500 μM